Learn More
Selleck Chemical LLC Labetalone hydrochloride S5088-25mg
SDP
Supplier:  Selleck Chemical LLC S508825MG
Encompass_Preferred
Labetalone hydrochloride (Benzoic amide) is hydrochloride form of labetalone a blocker of both alpha- and beta-adrenergic receptors that is used as an antihypertensive
Catalog No. 50-311-2235