Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Labetalone (hydrochloride) | 96441-14-4 | 99.3% | 1 ML
SDP

Catalog No. 5000339764
Encompass_Preferred

Labetalone hydrochloride is an impurity of Labetalol, which functions as an orally active adrenoceptor blocking agent by acting as a competitive antagonist at both alpha- and beta-adrenoceptor sites. It is suitable for use in laboratory research and the manufacture of substances.

  • An impurity of Labetalol
  • Acts as an orally active adrenoceptor blocking agent
  • Functions as a competitive antagonist at alpha- and beta-adrenoceptor sites
  • Molecular weight: 362.85
  • Chemical formula: C19H23ClN2O3
  • Purity: 99.3%
  • Appearance: Off-white to light yellow solid
  • Identified uses: Laboratory chemicals, manufacture of substances

Catalog No. 50-003-39764 Supplier Medchemexpress LLC Supplier No. HY12837910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Labetalone hydrochloride is an impurity of Labetalol, which functions as an orally active adrenoceptor blocking agent by acting as a competitive antagonist at both alpha- and beta-adrenoceptor sites. It is suitable for use in laboratory research and the manufacture of substances.

  • An impurity of Labetalol
  • Acts as an orally active adrenoceptor blocking agent
  • Functions as a competitive antagonist at alpha- and beta-adrenoceptor sites
  • Molecular weight: 362.85
  • Chemical formula: C19H23ClN2O3
  • Purity: 99.3%
  • Appearance: Off-white to light yellow solid
  • Identified uses: Laboratory chemicals, manufacture of substances

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.