Learn More
Neutec Group LabFlame Basic Safety Bunsen Burner
SDP
Catalog No. 14910408 Shop All Neutec Group Products
Change view
Click to view available options
Material:
Stainless Steel, Glass
Catalog No. Material
14-910-408 Stainless Steel, Glass
1 options
Catalog No. 14-910-408 Supplier Neutec Group Supplier No. BTLF8.201.000
Add to Cart
Add to Cart
Optimized for use in microbiology laboratories
The new Biotool Swiss LabFlame Safety Bunsen Burner has stainless steel housing, made of one piece free of any gaps or rims and therefore easy to clean and fully UV resistant. The optimized air-flow system allows uniform and constant flame even in safety cabinets with horizontal or vertical air flow.
Specifications
Material Stainless Steel, Glass
Description Dimensions: 130 x 103 x 49 mm (5.1 x 4.1 x 1.9 in.)
Max. Temperature (Metric) 1350°C (Liquid gas), 1300°C (Natural gas)
Product Type Safety Bunsen Burner with Foot Switch