Learn More
Cole-Parmer Cole-Parmer LabGEN Plastic Tip Probes, 0.25 to 50 mL, Hard Tissue, 25/pack
Catalog No. NC0189151 Shop All Cole-Parmer Products
Encompass_Preferred
These plastic tip probes are maintenance-free and do not require the disassembly associated with traditional stainless steel generators. Choose the generator or probe based on the dispersion or tissue type, and sample size.
Catalog No. NC0189151 Supplier Cole-Parmer Supplier No. 0472784
These plastic tip probes are maintenance-free and do not require the disassembly associated with traditional stainless steel generators. Choose the generator or probe based on the dispersion or tissue type, and sample size.