Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Minigrip™ LabGuard™ Biohazard Specimen Bags

Catalog No. 22311203
Change view
Click to view available options
Height (Metric):
22.86 cm
38.10 cm
Width (Metric):
15.2 cm
30.48 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric) Width (Metric)
22-311-203 22.86 cm 15.2 cm
22-311-205 38.10 cm 30.48 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-311-203 Supplier Minigrip™ Supplier No. SBL2AP69B
Add to Cart
Add to Cart

Provide safe and secure transport of multiple samples

  • Three walls
  • With reclosable zipper
  • Thickness of 1.75 mil

Specifications

Closure Type Ziplock
Color Clear
For Use With (Application) Provide safe and secure transport of multiple samples
Includes Absorbent pad, reclosable zipper, biohazard symbol and document pouch
Material Plastic
Quantity 50 Pack
Thickness (English) 1.75 mil
Height (Metric) 22.86 cm
Product Type 3-wall Biohazard Specimen Bag
Width (Metric) 15.2 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.