Learn More
Labjack Corporation LabJack T7 - for ultimate power and data acquisition flexibility
SDP
Catalog No. NC3357519
Encompass
Choose the LabJack T7 for ultimate power and data acquisition flexibility. With 16-bit analog inputs, the option to use USB or Ethernet, and numerous built-in firmware features, the T7 can help you accomplish nearly any DAQ need. The LabJack T7 also has 2 analog outputs (12-bit), 23 digital I/O, and up to 10 digital counters/timers.
Catalog No. NC3357519 Supplier Labjack Corporation Supplier No. T7
Choose the LabJack T7 for ultimate power and data acquisition flexibility. With 16-bit analog inputs, the option to use USB or Ethernet, and numerous built-in firmware features, the T7 can help you accomplish nearly any DAQ need. The LabJack T7 also has 2 analog outputs (12-bit), 23 digital I/O, and up to 10 digital counters/timers.