Learn More
Med Vet International Dynarex Disposable Labjacket w/Pockets, Large, White, 30/Case
Catalog No. 503047503
Encompass_Preferred
Dynarex Disposable Lab Jackets are designed to protect skin and clothes from light dirt, splashes, and spills. Constructed from breathable polypropylene material, these durable garments provide a comfortable, non-constricting fit over everyday clothing. Triple-layered polymer material resists moisture, dirt, and spills Waist-length garment provides ample upper-body coverage Elastic wrists provide a secure fit and reduce the risk of snags and tears Buttons in front for reliable closure Reuse or dispose as needed Not made with natural rubber latex Semi-transparent
Catalog No. 50-304-7503 Supplier Med Vet International Supplier No. 2004
Dynarex Disposable Lab Jackets are designed to protect skin and clothes from light dirt, splashes, and spills. Constructed from breathable polypropylene material, these durable garments provide a comfortable, non-constricting fit over everyday clothing. Triple-layered polymer material resists moisture, dirt, and spills Waist-length garment provides ample upper-body coverage Elastic wrists provide a secure fit and reduce the risk of snags and tears Buttons in front for reliable closure Reuse or dispose as needed Not made with natural rubber latex Semi-transparent