Learn More
Ace Glass, Inc. Lab jaws clamp holder, 90°, 18mm (0.71in) grip, Troemner916051
SDP
Catalog No. NC0091690
Encompass
18mm nickel-plated zinc, 90 degree frame clamp holder for use with all Labjaws extension clamps. Features a 0.71 grip. Troemner part number is 916051.
Catalog No. NC0091690 Supplier Ace Glass, Inc. Supplier No. 1108019
18mm nickel-plated zinc, 90 degree frame clamp holder for use with all Labjaws extension clamps. Features a 0.71 grip. Troemner part number is 916051.