Learn More
Ace Glass, Inc. Lab jaws open ring support, 4in, 2.28in arm, Troemner 916209
SDP
Catalog No. 50977218
Encompass
4 aluminum Labjaws open ring support with 5.7cm (2.28) arm is IDeal for use with funnels and boiling flasks.
Catalog No. 50-977-218 Supplier Ace Glass, Inc. Supplier No. 1117117
4 aluminum Labjaws open ring support with 5.7cm (2.28) arm is IDeal for use with funnels and boiling flasks.