Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS LabKlenz™ 100 Alkaline Detergent

Catalog No. 17463003 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
30 gal.
250 gal.
Container Type:
Drum
Plastic Bottles
Plastic jerrican
Tote
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Container Type
17-463-003 1 gal. Plastic Bottles
17-463-041 5 gal. Plastic jerrican
17-463-042 30 gal. Drum
17-463-043 250 gal. Tote
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-463-003 Supplier STERIS Supplier No. 1L1008
Add to Cart
Add to Cart

Alkaline detergent with corrosion inhibitors

  • Formulated to meet unique cleaning and materials compatibility requirements found in laboratory glassware washing applications
  • High alkaline for effective removal of residues in both soft and hard water
  • Contains corrosion inhibitors for excellent materials compatibility
  • Reduces utility consumption and cleaning time with low foaming, free-rinsing formulation
  • Phosphate-free formulation with biodegradable surfactants
  • Manufactured in cGMP compliant, FDA registered and ISO 9001 certified facility
  • Specific gravity: 1.15

Specifications

Appearance Yellow
Container Type Plastic Bottles
Quantity 4 Pk.
Volume (English) 1 gal.
Volume (Metric) 3.785 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.