Learn More
Research Products International Corp Labmat Protective Covering, 20 Inches Wide x 50 Feet Long per Roll
SDP
Catalog No. 50476629
Encompass
Labmat simplifies clean-up and protects surfaces from stains and spills. Flexible polyethylene film backing prevents solutions from soaking through. The highly absorbent top surface traps water, dyes, corrosive chemicals and radioactive solutions. The entire sheet can be curled up and discarded after contamination. Rolls are easily cut with scissors while roll is retained in dispenser.
Catalog No. 50-476-629 Supplier Research Products International Corp Supplier No. F24675
Labmat simplifies clean-up and protects surfaces from stains and spills. Flexible polyethylene film backing prevents solutions from soaking through. The highly absorbent top surface traps water, dyes, corrosive chemicals and radioactive solutions. The entire sheet can be curled up and discarded after contamination. Rolls are easily cut with scissors while roll is retained in dispenser.