Learn More
Research Products International Corp Labmat Sheets, 20 x 24 Inches, 100 Sheets per Case
SDP
Catalog No. 50213319
Encompass
Labmat simplifies clean-up and protects surfaces from stains and spills. Flexible polyethylene film backing prevents solutions from soaking through. The highly absorbent top surface traps water, dyes, corrosive chemicals and radioactive solutions. The entire sheet can be curled up and discarded after contamination.
Catalog No. 50-213-319 Supplier Research Products International Corp Supplier No. F246750020
Add to Cart
Add to Cart
Labmat simplifies clean-up and protects surfaces from stains and spills. Flexible polyethylene film backing prevents solutions from soaking through. The highly absorbent top surface traps water, dyes, corrosive chemicals and radioactive solutions. The entire sheet can be curled up and discarded after contamination.