Learn More
Alkali Scientific Labnet Electronic Pipette, Programmable, 12 Channel, 20uL
SDP
Catalog No. 5000662797
Encompass_Preferred
Labnet Electronic Pipette, Programmable • Utilize everyday TRADITIONAL TIPS • No need for Combitips • Highly accurate & Factory calibrated • Easy to use 4 button operation • Large LCD screen and simple interface • Ergonomically designed handle and finger rest • Lower portion easily removed for autoclaving • Rechargeable long life Li ion battery • 1 year warranty • Perfect for high volume qPCR and Real Time assays within Diagnostic Laboratories • Free up technician time, "knocks out" 96 well plates in record time. • Requires just a small movement of the index finger to mix, dispense & aspirate which reduces the risk of Repetitive Strain Injuries (RSI)
Catalog No. 50-006-62797 Supplier Alkali Scientific Supplier No. P3612L20
Labnet Electronic Pipette, Programmable • Utilize everyday TRADITIONAL TIPS • No need for Combitips • Highly accurate & Factory calibrated • Easy to use 4 button operation • Large LCD screen and simple interface • Ergonomically designed handle and finger rest • Lower portion easily removed for autoclaving • Rechargeable long life Li ion battery • 1 year warranty • Perfect for high volume qPCR and Real Time assays within Diagnostic Laboratories • Free up technician time, "knocks out" 96 well plates in record time. • Requires just a small movement of the index finger to mix, dispense & aspirate which reduces the risk of Repetitive Strain Injuries (RSI)