Learn More
Pipette Supplies, Inc. LABNET FLTR,SNGL CHNL,5ML,10ML
SDP
Catalog No. NC0983867
Encompass
Labnet Filter, Single Channel, 5ML, 10ML - 1pc A single filter for the following pipettes: Alphapette, Beta-Pette, BioPette A, BioPette Plus, Labpette Discovery, Ultra Ezpette and XL3000i
Catalog No. NC0983867 Supplier Pipette Supplies, Inc. Supplier No. LN SP19454
Add to Cart
Add to Cart
Labnet Filter, Single Channel, 5ML, 10ML - 1pc A single filter for the following pipettes: Alphapette, Beta-Pette, BioPette A, BioPette Plus, Labpette Discovery, Ultra Ezpette and XL3000i