Learn More
Labnet International, Inc Labnet Mini Centrifuge, Purple Lid
Supplier:  Labnet International, Inc C1601P
Encompass_Preferred
Simple and convenient to operate, the new Labnet mini microcentrifuges feature greater capacity (8-place MCT and 4-place PCR strip rotors), a quick-release rotor system and electronic brake. Once samples are loaded and the lid is closed, the rotor rapidly accelerates to 6,000 rpm, making it an ideal centrifuge for bringing small droplets to the bottom of tubes for micro-filtrations or basic separations.
Catalog No. NC1478094