Learn More
Sigma Aldrich Fine Chemicals Biosciences Labnet MultiGeneTM OptiMax1EA
Catalog No. 502909095
Encompass
The new MultiGene OptiMax brings new speed and features to our Thermal Cycler line. It utilizes an easy user interface having a much greater speed of operation by increasing the ramping speed and eliminates overshooting or undershooting with contributes to longer run times.New features like PC Viewer and a TM calculator allows you to connect to a Windows based PC using a USB interface and view the actual temperature profiles in real time. The new 6-block gradient format allows you to select the temperatures you wish to optimize rather than have the system select for you..nbspNo condensation after overnight cooling at 4C.nbsp200 complete programs in memory.nbsp6-segment block temperature control with users able to select 6-segment temps independently.nbsp6 temperature blocks in 4 x 4 well format.nbspPeltier heating/cooling method.nbspTM Calculator.nbspPC Viewer.nbspSimple user interface.nbspPower cables for either the UK 3-pin or European 2-pin plugs supplied
Catalog No. 50-290-9095 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7591551EA
The new MultiGene OptiMax brings new speed and features to our Thermal Cycler line. It utilizes an easy user interface having a much greater speed of operation by increasing the ramping speed and eliminates overshooting or undershooting with contributes to longer run times.New features like PC Viewer and a TM calculator allows you to connect to a Windows based PC using a USB interface and view the actual temperature profiles in real time. The new 6-block gradient format allows you to select the temperatures you wish to optimize rather than have the system select for you..nbspNo condensation after overnight cooling at 4C.nbsp200 complete programs in memory.nbsp6-segment block temperature control with users able to select 6-segment temps independently.nbsp6 temperature blocks in 4 x 4 well format.nbspPeltier heating/cooling method.nbspTM Calculator.nbspPC Viewer.nbspSimple user interface.nbspPower cables for either the UK 3-pin or European 2-pin plugs supplied