Learn More
Sigma Aldrich Fine Chemicals Biosciences Labnet PrismTM R Refrigera1EA
Catalog No. 502908675
Encompass
.nbspMax. Speed 13500 rpm.nbspMax. RCF 17 135 x g.nbspTemp. Range -10 to 40C.nbspDimensions (in.) 10.9(W) x 17.75(D) x 9.75(H).nbspDimensions (cm) 27.7(W) x 45(D) x 24.75(H).nbspTimer 0.5 to 99 min. with continuous and momentary.nbspCooling Time (Room Temp. to 4C) 8 min..nbspWeight 44 lb/20 kg
Catalog No. 50-290-8675 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7237621EA
Add to Cart
Add to Cart
Request a Quote
.nbspMax. Speed 13500 rpm.nbspMax. RCF 17 135 x g.nbspTemp. Range -10 to 40C.nbspDimensions (in.) 10.9(W) x 17.75(D) x 9.75(H).nbspDimensions (cm) 27.7(W) x 45(D) x 24.75(H).nbspTimer 0.5 to 99 min. with continuous and momentary.nbspCooling Time (Room Temp. to 4C) 8 min..nbspWeight 44 lb/20 kg