Learn More
LABOMED™ Lx 300 LED Laboratory Microscope
SCI_ED
Catalog No. S37242
Change view
Click to view available options
Head Style:
Binocular
Catalog No. Head Style
S37242 Binocular
1 options
Catalog No. S37242 Supplier Labo America Inc Supplier No. 9136001
Add to Cart
Add to Cart
Mobile and easy to use. LABOMED™ Lx 300 LED Laboratory Microscope features comfortable viewing angle, light weight construction, and built-in LED LioN illumination system.
Stand:
  • Single mold sturdy stand with anti-rust materials
  • Ergonomic base for hand rest and greater stability
Viewing Body:
  • Siedentopf Binocular (Butterfly style) or Trinocular head
  • 45° inclined, 360° rotatable, with adjustable viewing height up to 40mm
  • IPD: 48 - 75mm
Eyepieces:
  • Wide field 10x/18mm with foldable eye guards, lockable, antifungal
Nosepiece:
  • Reverse angle quadruple nosepiece (Ball bearing type) with click stops and rubber grip
Objectives:
  • EP2 Plan Achromat DIN Infinity objectives 4x, 10x, 40x (spring loaded), 100x (spring loaded, oil), antifungal
Mechanical stage:
  • Double plate stage 150 x 135mm, X-Y travel range 75mm x 40mm with low-height controls
  • Hard coated surface for scratch resistance
  • Double specimen holder
Condenser:
  • Sub stage Abbe condenser NA 1.25 with aspheric lens
  • Iris diaphragm with blue daylight filter
  • Rack and pinion movements on metal guides
Focusing:
  • Coaxial coarse and fine focus on ball drive system for smooth operation
Illumination:
  • Intensity controlled LED (up to 50,000 hours of LED life)
Electrical requirements:
  • Universal input 100V - 240V AC, 50/60Hz
  • Built-in voltage stabilizer
Optional Accessories:
  • Dark field kit, phase contrast kit, video adapter, digital camera and mirror attachment
Certifications
CE/UL
Warranty and Services
5 years parts and 1 year electrical
Specifications
Condenser Type 1.25 NA Abbe condenser
Eyepiece 10X/18 mm with Foldable Eyeguard
Head Style Binocular
Lighting Type LED
Magnification Power 4X, 10X, 40X, 100X
Model Lx 300
Product Type Microscope
Resolution Coarse and fine
For Use With (Application) Education, Biology, Laboratory
Includes Power cable (6 ft.), Cargille immersion oil, Spare bulb (20w version only), Nylon dust cover and cleaning cloth, Styrofoam casing
Nosepiece Reverse angle quadruple nosepiece
Electrical Requirements 100/240 V 50/60 Hz