Learn More
BURKLE INC Scoop LaboPlast Bio, single-use, Bio-PE, 0.85 oz. (25 ml)
Catalog No. 503081197
Encompass_Preferred
Scoop LaboPlast Bio, single-use, made from sustainable green PE bio-plastic. This innovative Bio-PE material combines the proven properties of conventional polyethylene — durability, chemical resistance, and food safety — with an eco-friendly manufacturing process using renewable raw materials. Fully recyclable for a reduced environmental footprint. Perfect for hygienic, contamination-free sampling in laboratories, food production, pharmaceuticals, and environmental analysis. Capacity: 25 ml (0.85 oz).
Catalog No. 50-308-1197 Supplier BURKLE INC Supplier No. 53790001
Scoop LaboPlast Bio, single-use, made from sustainable green PE bio-plastic. This innovative Bio-PE material combines the proven properties of conventional polyethylene — durability, chemical resistance, and food safety — with an eco-friendly manufacturing process using renewable raw materials. Fully recyclable for a reduced environmental footprint. Perfect for hygienic, contamination-free sampling in laboratories, food production, pharmaceuticals, and environmental analysis. Capacity: 25 ml (0.85 oz).