Learn More
BURKLE INC Scoop LaboPlast Bio, single-use, Bio-PE, 1.69 oz. (50 ml)
Catalog No. 503081198
Encompass_Preferred
Scoop LaboPlast Bio, single-use, made from eco-friendly green PE bio-plastic. The innovative Bio-PE material delivers the same durability, chemical resistance, and food safety as conventional polyethylene, but is produced from renewable raw materials and is fully recyclable. Designed for hygienic, contamination-free sampling in laboratories, food production, pharmaceuticals, and environmental testing. Capacity: 50 ml (1.69 oz).
Catalog No. 50-308-1198 Supplier BURKLE INC Supplier No. 53790003
Scoop LaboPlast Bio, single-use, made from eco-friendly green PE bio-plastic. The innovative Bio-PE material delivers the same durability, chemical resistance, and food safety as conventional polyethylene, but is produced from renewable raw materials and is fully recyclable. Designed for hygienic, contamination-free sampling in laboratories, food production, pharmaceuticals, and environmental testing. Capacity: 50 ml (1.69 oz).