Learn More
BURKLE INC Scoop LaboPlast Bio, single-use, Bio-PE, 3.38 oz. (100 ml)
Catalog No. 503081272
Encompass_Preferred
Scoop LaboPlast Bio, single-use, made from sustainable green PE bio-plastic. This innovative Bio-PE material offers all the advantages of conventional polyethylene — such as durability, chemical resistance, and food safety — but is produced from renewable raw materials and is 100% recyclable. Ideal for hygienic, contamination-free sampling in laboratories, food production, pharmaceuticals, and environmental testing. Capacity: 100 ml (3.38 oz).
Catalog No. 50-308-1272 Supplier BURKLE INC Supplier No. 53790005
Scoop LaboPlast Bio, single-use, made from sustainable green PE bio-plastic. This innovative Bio-PE material offers all the advantages of conventional polyethylene — such as durability, chemical resistance, and food safety — but is produced from renewable raw materials and is 100% recyclable. Ideal for hygienic, contamination-free sampling in laboratories, food production, pharmaceuticals, and environmental testing. Capacity: 100 ml (3.38 oz).