Learn More
BURKLE INC Scoop LaboPlast Bio, single-use, Bio-PE, 5.07 oz. (150 ml)
Catalog No. 503081364
Encompass_Preferred
Scoop LaboPlast Bio, single-use, made from sustainable green PE bio-plastic. This Bio-PE material offers the durability, chemical resistance, and food safety of conventional polyethylene while being manufactured from renewable raw materials and fully recyclable. Ideal for hygienic and contamination-free sampling in laboratories, food production, pharmaceuticals, and environmental testing. Capacity: 150 ml (5.07 oz).
Catalog No. 50-308-1364 Supplier BURKLE INC Supplier No. 53790007
Scoop LaboPlast Bio, single-use, made from sustainable green PE bio-plastic. This Bio-PE material offers the durability, chemical resistance, and food safety of conventional polyethylene while being manufactured from renewable raw materials and fully recyclable. Ideal for hygienic and contamination-free sampling in laboratories, food production, pharmaceuticals, and environmental testing. Capacity: 150 ml (5.07 oz).