Learn More
BURKLE INC Spoon LaboPlast single-use, PS, 0.085 oz (2.5 ml)
Catalog No. 503081234
Encompass_Preferred
The disposable spoon made of polystyrene is produced in our clean room of class 7 (10,000) and individually packaged for disposable use.
The spoon is available in two sizes, 0.08 oz. (2.5 ml) (0.08 oz.) (similar to a teaspoon) and 10 ml (0.34 oz.) (similar to a tablespoon), and is ideal for sampling powders, granulates and fluids.
Catalog No. 50-308-1234 Supplier BURKLE INC Supplier No. 53780011
The disposable spoon made of polystyrene is produced in our clean room of class 7 (10,000) and individually packaged for disposable use.
The spoon is available in two sizes, 0.08 oz. (2.5 ml) (0.08 oz.) (similar to a teaspoon) and 10 ml (0.34 oz.) (similar to a tablespoon), and is ideal for sampling powders, granulates and fluids.