Learn More
Sigma Aldrich Fine Chemicals Biosciences Laboratory brushes flask L
Catalog No. 502901380
Encompass
Brush materials resistant to acids aromatic and aliphatic hydrocarbons ketones ethyl acetate esters trichloroethylene and hot water. fDA and USDA approved materials.
Catalog No. 50-290-1380 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2813441PAK
Brush materials resistant to acids aromatic and aliphatic hydrocarbons ketones ethyl acetate esters trichloroethylene and hot water. fDA and USDA approved materials.