Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Laboratory Chemical Spill Kit

Catalog No. 17111299 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
13.8 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-299 13.8 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-299 Supplier Brady™ Supplier No. SPCFSHPROMO

Contains all the products needed to support acid, caustic, halogen and solvent spill control needs, all in one convenient kit.

Ideal for labs, hospitals and other areas where chemicals are prevalent. Provides quick response and ease of disposal, these pads and rolls support a clean, efficient work environment.

Kit includes:
  • Fisher Cat 19-140-740: (100) 7.5 x 10 in. chemical absorbent pads - perfect for smaller leaks, drips and spills and easy bench top access
  • Fisher Cat  19-140-743: (1) 7.5 in. x 50 ft. chemical absorbent roll - conveniently perforated to allow you to use exactly what you need
  • Fisher Cat 19-150-268: (1) Chemical spill kit designed to absorb up to 5 gallons of liquid to support larger spill clean-up and conveniently hang on the wall or fit under your fume hood.
  • (10) 15 x 19 in. chemical absorbent pads
  • (2) 3 in. x 4 ft. absorbent socks
  • Disposal bag
  • Instructions sheet
  • Nitrile gloves

Specifications

Capacity (English) 13.8 gal.
Type Spill Control Kit
For Use With (Application) Acid, caustic, halogen and solvent spill control
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.