Learn More
Boca Scientific Inc Laboratory Columns, 2.5ml with 90um Inserted Lower Filter, Pack of 20
Catalog No. NC1894518
Encompass
These empty 2.5ml laboratory columns come with an inserted lower filter with a pore size of 90um. Each column has Luer-lock connectors on both sides for tubing and syringes. To close the columns tightly, the 2.5ml columns have a Luer-lock closing cap. These columns can be centrifuged and autoclaved (with filters at 110°C). Each package of columns includes two one-way stopcocks.
Catalog No. NC1894518 Supplier Boca Scientific Inc Supplier No. S10129
These empty 2.5ml laboratory columns come with an inserted lower filter with a pore size of 90um. Each column has Luer-lock connectors on both sides for tubing and syringes. To close the columns tightly, the 2.5ml columns have a Luer-lock closing cap. These columns can be centrifuged and autoclaved (with filters at 110°C). Each package of columns includes two one-way stopcocks.