Learn More
Boca Scientific Inc Laboratory Columns, 5ml with 35um Inserted Lower Filter, Pack of 20
Catalog No. NC1894522
Encompass
These empty 5ml laboratory columns come with an inserted lower filter with a pore size of 35um. Each column has Luer-lock connectors on both sides for tubing and syringes. To close the columns tightly, the 5ml columns include an additional cap. Each package of columns includes two one-way stopcocks.
Catalog No. NC1894522 Supplier Boca Scientific Inc Supplier No. S1013
These empty 5ml laboratory columns come with an inserted lower filter with a pore size of 35um. Each column has Luer-lock connectors on both sides for tubing and syringes. To close the columns tightly, the 5ml columns include an additional cap. Each package of columns includes two one-way stopcocks.