Learn More
Alkali Scientific ASI™ Laboratory Foam Bench, Drawer and Shelf Liner, 1/8"x30"x50', 1/Pack
SDP
Catalog No. 503028904
Encompass_Preferred
A resilient surface that helps to cushion and protect sensitive instruments. Can be easily cut to exact desired size. May be taped or otherwise secured in place. Both sides are usable.  Cross-linked, polyethylene, closed-cell foam Non-absorbent 2 lb. density Easy to clean No chloroflourocarbon blowing agents used
Catalog No. 50-302-8904 Supplier Alkali Scientific Supplier No. FL003
A resilient surface that helps to cushion and protect sensitive instruments. Can be easily cut to exact desired size. May be taped or otherwise secured in place. Both sides are usable.  Cross-linked, polyethylene, closed-cell foam Non-absorbent 2 lb. density Easy to clean No chloroflourocarbon blowing agents used