Learn More
Research Products International Corp Compact Lab Incubator, 0.7 Cubic Feet Capacity
SDP
Catalog No. 50212159
Encompass
Versatile incubators designed for reliable and dependable laboratory use. Ideal for research, clinical and industrial laboratory applications. Accurate and uniform temperatures up to 62°C can be maintained. The sensitive heat controller regulates temperature accuracy to ±1/4°C. Advanced styling features include a see through tinted acrylic door to view contents. Cabinets have an attractive ivory enamel finish and aluminum interior. Illuminated switches indicate when power and heat cycles are on.
Catalog No. 50-212-159 Supplier Research Products International Corp Supplier No. 110140
Versatile incubators designed for reliable and dependable laboratory use. Ideal for research, clinical and industrial laboratory applications. Accurate and uniform temperatures up to 62°C can be maintained. The sensitive heat controller regulates temperature accuracy to ±1/4°C. Advanced styling features include a see through tinted acrylic door to view contents. Cabinets have an attractive ivory enamel finish and aluminum interior. Illuminated switches indicate when power and heat cycles are on.