Learn More
Research Products International Corp Compact Lab Incubator, 2.0 Cubic Feet Capacity
SDP
Catalog No. 50212163
Encompass
Versatile incubators designed for reliable and dependable laboratory use. Ideal for research, clinical and industrial laboratory applications. Accurate and uniform temperatures up to 62°C can be maintained. The sensitive heat controller regulates temperature accuracy to ±1/4°C. Advanced styling features include a see through tinted acrylic door to view contents. Cabinets have an attractive ivory enamel finish and aluminum interior. Illuminated switches indicate when power and heat cycles are on.
Supplied with chrome plated wire shelves, thermometer, three prong grounded plug and instruction manual. Large cabinet has two adjustable and one fixed shelves. Incubators are fully tested prior to shipment and are warranted for 18 months.
Usually ships within 3 weeks.
Catalog No. 50-212-163 Supplier Research Products International Corp Supplier No. 112140
Versatile incubators designed for reliable and dependable laboratory use. Ideal for research, clinical and industrial laboratory applications. Accurate and uniform temperatures up to 62°C can be maintained. The sensitive heat controller regulates temperature accuracy to ±1/4°C. Advanced styling features include a see through tinted acrylic door to view contents. Cabinets have an attractive ivory enamel finish and aluminum interior. Illuminated switches indicate when power and heat cycles are on.
Supplied with chrome plated wire shelves, thermometer, three prong grounded plug and instruction manual. Large cabinet has two adjustable and one fixed shelves. Incubators are fully tested prior to shipment and are warranted for 18 months.
Usually ships within 3 weeks.