Learn More
Sonics & Materials Inc LABORATORY JACK 6IN X 5 IN TOP
SDP
Catalog No. NC2588123
Encompass
Laboratory Jack 6in x 5 in topprovides adjustable elevation from64mm to 254mm 6in x 5 in top plate
Catalog No. NC2588123 Supplier Sonics & Materials Inc Supplier No. 83000113
Add to Cart
Add to Cart
Laboratory Jack 6in x 5 in topprovides adjustable elevation from64mm to 254mm 6in x 5 in top plate