Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Tuttnauer™ Lab Line ELC-PVG-P Autoclave
SDP

Catalog No. 13890053 Shop All Heidolph North America Products
Change view
Click to view available options
Capacity (Metric):
28 L
85 L
160 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
13-890-053 85 L
13-890-050 28 L
13-890-052 160 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 13-890-053 Supplier Heidolph™ Supplier No. 023210678
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Autoclave offers a steam generator, vacuum pump, and cooling coils for efficient air removal and complete drying. Designed to sterilize glassware, plastics, liquids and media.

Standard Operating Features:

  • Designed to provide high quality, repeatable performance and accountability for a wide range of applications used in modern laboratories
  • For use with Agar preparation as well as the sterilization of liquids, pipettes, glasses, instruments, and biohazards with various cooling options
  • Equipped with a vacuum pump, cooling coils, and a steam generator for all laboratory sterilization needs
  • Compact size allows autoclave to fit in your lab.
  • Fast cooling (C) saves up to 75% of the cool down times of liquid samples
  • Immediate Steam (G) and Efficient Air Removal (PV) Steam generator in combination with vacuum (PV G) for complete drying
  • PID (Proportional Integral Differential) pressure control
  • Stores the last 200 cycles in built-in memory
  • Two independent, flexible PT100 temperature sensors help prevent boil over of liquids
  • Compressed air is passed through a microbiological filter into the autoclave chamber after sterilization to prevent a drop in pressure which causes load deformation, cracks, and spills
  • Cold water is circulated through cooling pipes to rapidly reduce the temperature of the chamber and liquid load to a safe temperature
  • Highly efficient drying is achieved by uniformly heating the chamber wall of the benchtop autoclave using a coiled pipe
  • The vacuum pump can be used for post vacuum drying to ensure improved drying of porous loads and hollow instruments such as pipette tips
  • Double locking safety device prevents door from opening while chamber is pressurized
  • Control lock-out switch prevents starting a cycle if door is not properly locked
  • Control system protection prevents door from opening at high pressure and temperature
  • All units include trays and printer
  • Units require utility connection to treated water, tap water, compressed air, and a floor sink

Specifications

Autoclavable Not Autoclavable
Amperage ∼16 A
Certifications/Compliance UL, cUL, ASME
Depth (Metric) Tray 67 cm
Depth (English) Tray 26 in.
Capacity (English) 22 gal.
Capacity (Metric) 85 L
Depth (Metric) Chamber 70 cm
Diameter (English) Chamber 15 in.
Depth (English) Chamber 28 in.
Diameter (Metric) Chamber 38 cm
For Use With (Application) Sterilization
For Use With (Equipment) Liquids, Glassware, Utensils (Wrapped or Unwrapped), Hollow Items, and Solids
Height (English) Exterior 53.8 in.
Height (Metric) Exterior 136.6 cm
Model 3870 EL-PV-GC
Product Type Autoclave
Temperature (English) 221°F to 278°F
Temperature (Metric) 105°C to 137°C
Display Multi-color to display the stage of the cycle
Net Weight (English) 286.6 lb.
Net Weight (Metric) 130 kg
Printer Included
Voltage 230 V
Wattage 4800 w
Width (English) Tray 14 in.
Width (Metric) Tray 35 cm
Sterility Non-sterile
Shipping Weight (Metric) 102 kg
Dimensions (L x W x H) 28.3 x 37 x 21.3 in. (72 x 94 x 54 cm)
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.