Learn More
Research Products International Corp Laboratory Marker Pen, Black Ink
SDP
Catalog No. 50550175
Encompass
This unique Laboratory Marker Pen is ideal for marking and identifying a wide variety of items. Features a dual tip design with a 0.7 mm micro-fine tip and 1.2 mm fine tip. Marks on any surface including glass, plastic, metal and paper. After drying (20 - 30 seconds) markings become waterproof and highly resistant to solvents and alcohols. Markings remain legible at low temperature storage down to -70°C.
Catalog No. 50-550-175 Supplier Research Products International Corp Supplier No. 145150
This unique Laboratory Marker Pen is ideal for marking and identifying a wide variety of items. Features a dual tip design with a 0.7 mm micro-fine tip and 1.2 mm fine tip. Marks on any surface including glass, plastic, metal and paper. After drying (20 - 30 seconds) markings become waterproof and highly resistant to solvents and alcohols. Markings remain legible at low temperature storage down to -70°C.