Learn More
Research Products International Corp Laboratory Gravity Convection Oven, Hydraulic Thermostat, 750 Watts
SDP
Catalog No. 50476430
Encompass
Gravity Convection Oven, 1.27 Cubic Feet
Designed for years of dependable service. Well-crafted and versatile, they are used for part drying, baking, curing, sterilizing, evaporating, heat treating, annealing, and testing. Cabinet exteriors are constructed of heavy steel with a hard scratch resistant hammer finish. Aluminized steel interiors are corrosion resistant. Energy efficient heating elements are designed for quick run-up and recovery times and provide excellent temperature uniformity. Ovens have a temperature range to 450°F / 232°C. Illuminated indicator lamps and two adjustable wire racks are included.
WARNING  This product can expose you to chemicals including Diethylhexyl Phthalate (DEHP), which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-476-430 Supplier Research Products International Corp Supplier No. 20270
Gravity Convection Oven, 1.27 Cubic Feet
Designed for years of dependable service. Well-crafted and versatile, they are used for part drying, baking, curing, sterilizing, evaporating, heat treating, annealing, and testing. Cabinet exteriors are constructed of heavy steel with a hard scratch resistant hammer finish. Aluminized steel interiors are corrosion resistant. Energy efficient heating elements are designed for quick run-up and recovery times and provide excellent temperature uniformity. Ovens have a temperature range to 450°F / 232°C. Illuminated indicator lamps and two adjustable wire racks are included.
WARNING  This product can expose you to chemicals including Diethylhexyl Phthalate (DEHP), which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.