Learn More
PHCbi 23.7 cu. ft. Upright Laboratory Refrigerator
Catalog No. 502720043
Encompass_Preferred
The MPR-722R-PA is an upright pharmacy refrigerator offering a stable and reliable environment for managing clinical, pharmaceutical and industrial research at uniform temperatures. Easy to use features and design and precise temperature control provides the ideal climate for various laboratory procedures including biomedical storage and processing.Effective capacity: 23.7 cu.ft. (671 L)Temperature range: 2℃ to 23℃
Catalog No. 50-272-0043 Supplier PHCbi Supplier No. MPR722RPA
Add to Cart
Add to Cart
Request a Quote
The MPR-722R-PA is an upright pharmacy refrigerator offering a stable and reliable environment for managing clinical, pharmaceutical and industrial research at uniform temperatures. Easy to use features and design and precise temperature control provides the ideal climate for various laboratory procedures including biomedical storage and processing.Effective capacity: 23.7 cu.ft. (671 L)Temperature range: 2℃ to 23℃