Learn More
PHCbi 48.0 cu.ft. Large Capacity Laboratory Refrigerator
Catalog No. 502720045
Encompass_Preferred
The MPR-1412R-PA is a pharmacy grade refrigerator delivering a stable and reliable environment for managing clinical, pharmaceutical and biomedical storage and processing at uniform temperatures. Easy to use features and design and precise temperature control with fast recovery following door openings, assures safe storage during frequent access and high ambient conditions.Effective capacity: 48.0 cu.ft. (1359 L)Temperature range: 2℃ to 23℃
Catalog No. 50-272-0045 Supplier PHCbi Supplier No. MPR1412RPA
Add to Cart
Add to Cart
Request a Quote
The MPR-1412R-PA is a pharmacy grade refrigerator delivering a stable and reliable environment for managing clinical, pharmaceutical and biomedical storage and processing at uniform temperatures. Easy to use features and design and precise temperature control with fast recovery following door openings, assures safe storage during frequent access and high ambient conditions.Effective capacity: 48.0 cu.ft. (1359 L)Temperature range: 2℃ to 23℃