Learn More
Cole-Parmer Cole-Parmer Laboratory Sample Labeling Kit
Supplier:  Cole-Parmer 2500031
Encompass_Preferred
Convenient kit includes twenty-four rolls of tape and six black markersSave money and color code your samples — tape comes in assorted colors. Durable labeling tape is waterproof, and oil and acid resistant. Each roll is 14 yards (12.8 m) long and 1/2" (12.7 mm) wide, and withstands temperatures from –10 to 250ºF (–23 to 121ºC). The permanent markers are also waterproof, and the nonfading ink dries instantly. The extra-fine 0.3 mm writing tip stays sharp and precise.
Catalog No. NC3792673