Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laboratory Start-Up Kit

Catalog No. 02003305 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-003-305 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-003-305 Supplier Avantor Supplier No. 989901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Molecular Biology Kit

Contains:

  • Tris (base), 250g, CAS 77-86-1, Cat. No. 4109-04
  • Disodium EDTA, 100g, CAS 139-33-3, Cat. No. 4040-00
  • Boric Acid, 500g, CAS 10043-35-3, Cat. No. 4035-01
  • Glycine, 250g, CAS 56-40-6, Cat. No. 4059-00
  • Tris Hydrochloride, 250g, CAS 1185-53-1, Cat. No. 4103-04
  • Sodium Chloride, 500g, CAS 7647-14-5, Cat. No. 4058-01

Specifications

Chemical Name or Material Laboratory Start-Up Kit
Name Note Ultrapure Bioreagent
CAS 1185-53-1, 7647-14-5, 10043-35-3, 139-33-3, 77-86-1, 56-40-6
Quantity 1 Kit
Concentration or Composition (by Analyte or Components) Tris, Disodium EDTA, Boric Acid, Glycine, Tris Hydrochloride, Sodium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.