Learn More
Research Products International Corp Water Resistant Stopwatch
SDP
Catalog No. 50212759
Encompass
Counts up to 24 hours in 1/100th second resolution for the first 30 minutes with a 1 second resolution thereafter. Split interval function. Also has a 12/24 hour alarm clock, calendar, LCD display, and an hourly chime option.
5 Year Warranty
Catalog No. 50-212-759 Supplier Research Products International Corp Supplier No. 810012
Counts up to 24 hours in 1/100th second resolution for the first 30 minutes with a 1 second resolution thereafter. Split interval function. Also has a 12/24 hour alarm clock, calendar, LCD display, and an hourly chime option.
5 Year Warranty