Learn More
Research Products International Corp Laboratory Timer, 4 Channel, with Memory, 100 Hour Range, 1 Second Resolution
SDP
Catalog No. 50550352
Encompass
4 independent channels each of which counts up or down for any interval from 1 second to 100 hours, plus clock button. Extra large display indicates channels in use. Each one has a distinctive 1 minute alarm. Automatically begins to count up to indicate time elapsed after count down reaches zero. Memory recalls last timer setting or reuse. Also has a clock mode. Has a stand, pocket clip, and a magnet. Supplied with one button cell battery.
Catalog No. 50-550-352 Supplier Research Products International Corp Supplier No. 810015
4 independent channels each of which counts up or down for any interval from 1 second to 100 hours, plus clock button. Extra large display indicates channels in use. Each one has a distinctive 1 minute alarm. Automatically begins to count up to indicate time elapsed after count down reaches zero. Memory recalls last timer setting or reuse. Also has a clock mode. Has a stand, pocket clip, and a magnet. Supplied with one button cell battery.