Learn More
BURKLE INC Laboratory tray, PP white, in. LxW 11.81x15.75 in. (300x400mm), 3.96 gal. (15l)
Catalog No. 5000025803
Encompass_Preferred
The laboratory trays are practical all-purpose trays that can be nested when empty, also durable, shatterproof and resist flexing. The trays have a strong, rounded rim.
Suitable as a spill trough for environmentally dangerous, aggressive substances, for experiments, as an etching tray, as an instrument holder, a medicine tray, a photographic development basin, a goods presentation tray and a display tray for foodstuffs.
Catalog No. 50-000-25803 Supplier BURKLE INC Supplier No. 42013040
The laboratory trays are practical all-purpose trays that can be nested when empty, also durable, shatterproof and resist flexing. The trays have a strong, rounded rim.
Suitable as a spill trough for environmentally dangerous, aggressive substances, for experiments, as an etching tray, as an instrument holder, a medicine tray, a photographic development basin, a goods presentation tray and a display tray for foodstuffs.