Learn More
Boekel Scientific™ Laboratory Water Bath Cleaner
SDP
Catalog No. 04100563 Shop All Boekel Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Water Baths, Plasma Thawing Baths
Catalog No. For Use With (Equipment)
04-100-563 Water Baths, Plasma Thawing Baths
1 options
Catalog No. 04-100-563 Supplier Boekel Scientific™ Supplier No. B1906001
Add to Cart
Add to Cart
The Laboratory Water Bath Cleaner is an all-purpose biocide. Add 20 drops per gallon of this water bath additive once every two weeks to control organic particles and contamination.
  • Size: 8 fl oz.
Specifications
For Use With (Equipment) Water Baths, Plasma Thawing Baths
Type Water Additive