Description
- Manage identification and antibiotic susceptibility testing (ID/AST) data right from your laboratory workstation
- Improve productivity by decreasing trips to MicroScan instrumentation
- Access LabPro data processing even when the system's computer is being used for panel loading/unloading or maintenance activities
- Consolidate data from multiple testing systems for epidemiology and other management reports
- Utilize one LIS connection for multiple testing systems
Specifications
Specifications
| Type | Adapter |
| For Use With (Equipment) | Beckman Coulter Analyzers |
| Includes | LabPro Connect for Closed Systems |