Learn More
Wells Lamont Industrial PointBreak™ LabPro Gloves
Catalog No. 17033400 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Hand:
Left
Right
Specific
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
17-033-400 2X-Large Specific
17-033-383 X-Small Left
17-033-384 Small Left
17-033-385 Medium Left
17-033-386 Large Left
17-033-387 X-Large Left
17-033-388 2X-Large Left
17-033-389 X-Small Right
17-033-390 Small Right
17-033-391 Medium Right
17-033-392 Large Right
17-033-393 X-Large Right
17-033-394 2X-Large Right
17-033-395 X-Small Specific
17-033-396 Small Specific
17-033-397 Medium Specific
17-033-398 Large Specific
17-033-399 X-Large Specific
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 17-033-400 Supplier Wells Lamont Industrial Supplier No. PB100XXL
Add to Cart
Add to Cart
Level 5 needlestick resistant glove, a breakthrough in protection designed using new PointBreak™ Technology.
WLI PointBreak™ LabPro™ gloves offer advanced protection where it matters most - covering the first three fingers, part of the palm, and fully wrapping the first finger for defense against needlestick injuries. Perfect for delicate procedures like animal inoculation and handling, organ recovery, and needlestick capping. These gloves deliver unmatched safety without compromising on comfort or dexterity. With the highest level of needlestick resistance, WLI ensures that professionals can work confidently, knowing they’re protected by the best.
Note: The A9 cut resistance only applies to the puncture resistant material, which is on the first 3 fingers and thumb crotch area. The cut resistance is not on the whole glove.
Specifications
Construction Cut and Sewn
Coating Material Uncoated
Color Black
Cut Resistance Rating ANSI A9
Hand Specific
Latex Free Yes
Material Polyester
Product Type Industrial Gloves
Size 2X-Large
Gender Unisex
Glove Cuff Style Hemmed
Grip Design Smooth
Lining Lined
Packaging Type Pair
Puncture Rating ANSI/ISEA 105-16 Level 5
Thumb Style Wing