Learn More
Grainger Elga Labpure S3 Purification Cartridge, High Density Polyethylene, 1 ea
Catalog No. 502791432
Encompass_Preferred
The Labpure S3 Low Ionic Ultra Purification Cartridge is a high-density polyethylene cartridge designed to effectively remove ionic and organic contaminants from feed water. It is engineered to maintain high water quality and electrical resistance in specialized water purification systems.
  • Contains sulphonated copolymer of styrene and divinylbenzene in the hydrogen form
  • Ensures consistent removal of ionic contaminants to achieve specified water quality
  • Helps prevent the release of weakly bound ions into product water
  • Supports the long-term maintenance of water purification system integrity
Catalog No. 50-279-1432 Supplier Grainger Supplier No. 16D261
The Labpure S3 Low Ionic Ultra Purification Cartridge is a high-density polyethylene cartridge designed to effectively remove ionic and organic contaminants from feed water. It is engineered to maintain high water quality and electrical resistance in specialized water purification systems.
  • Contains sulphonated copolymer of styrene and divinylbenzene in the hydrogen form
  • Ensures consistent removal of ionic contaminants to achieve specified water quality
  • Helps prevent the release of weakly bound ions into product water
  • Supports the long-term maintenance of water purification system integrity