Learn More
Grainger Elga Labpure S5 Purification Cartridge, Mixed Bed Resin, 1 ea
Catalog No. 502791371
Encompass_Preferred
The Labpure S5 purification cartridge is designed for use with Classic MKII systems to effectively remove ionic contaminants and achieve high-purity water. This mixed bed resin cartridge supports diverse laboratory applications, ensuring consistent water quality through reliable deionization.
  • Provides high-purity type I water with 18.2 M-cm resistivity.
  • Removes ionic contaminants to support life science and analytical applications.
  • Capacity of 45,000 liters at 18 M-cm at pH 7.0 per pack.
  • Features mixed bed resin for efficient ion exchange performance.
  • Designed for pretreated feedwater sources including reverse osmosis, filtered service deionization, or distilled water.
  • Optimized for low-cost, high-efficiency water purification in a compact format.
Catalog No. 50-279-1371 Supplier Grainger Supplier No. 16D263
The Labpure S5 purification cartridge is designed for use with Classic MKII systems to effectively remove ionic contaminants and achieve high-purity water. This mixed bed resin cartridge supports diverse laboratory applications, ensuring consistent water quality through reliable deionization.
  • Provides high-purity type I water with 18.2 M-cm resistivity.
  • Removes ionic contaminants to support life science and analytical applications.
  • Capacity of 45,000 liters at 18 M-cm at pH 7.0 per pack.
  • Features mixed bed resin for efficient ion exchange performance.
  • Designed for pretreated feedwater sources including reverse osmosis, filtered service deionization, or distilled water.
  • Optimized for low-cost, high-efficiency water purification in a compact format.