Learn More
Fine Science Tools LaGrange Scissors, Curved, Stainless Steel, 11.5 cm
SDP
Catalog No. NC9903044
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 18mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: Yes
Catalog No. NC9903044 Supplier Fine Science Tools Supplier No. 1417312
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 18mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: Yes