Learn More
Labrepco Inc LABREPCO 8.1 Cu.Ft. CO2 Incubator, Direct Heat & Air Jacket System, USB Data Logging, 8.1 cu.ft.
SDP
Catalog No. NC3737946
Encompass_Preferred
The LABREPCO 8.1 cu.ft. CO2 Incubator offers precise control for cell culture environments, essential for high-throughput applications. Its innovative heating system ensures stable temperature and CO2 levels, while advanced decontamination options enhance safety. A user-friendly touchscreen and USB data logging set it apart, making it ideal for labs requiring reliability in experimentation.
Catalog No. NC3737946 Supplier Labrepco Inc Supplier No. MCO230AICUVHLPA
Add to Cart
Add to Cart
Request a Quote
The LABREPCO 8.1 cu.ft. CO2 Incubator offers precise control for cell culture environments, essential for high-throughput applications. Its innovative heating system ensures stable temperature and CO2 levels, while advanced decontamination options enhance safety. A user-friendly touchscreen and USB data logging set it apart, making it ideal for labs requiring reliability in experimentation.