Learn More
Labrepco Inc LABREPCO FREEZER RACKS
SDP
Catalog No. NC1815812
Encompass_Preferred
Vertical Freezer Racks for 3.75″ High Boxes- Rack Only – 5 Boxes High Vertical freezer racks for Chest Freezers and Liquid Nitrogen Freezers. Freezer Racks are made of corrosion resistant stainless steel and include stainless steel locking rods. Security lock devices are also available for each rack though ordered separately.
Catalog No. NC1815812 Supplier Labrepco Inc Supplier No. CF5375
Vertical Freezer Racks for 3.75″ High Boxes- Rack Only – 5 Boxes High Vertical freezer racks for Chest Freezers and Liquid Nitrogen Freezers. Freezer Racks are made of corrosion resistant stainless steel and include stainless steel locking rods. Security lock devices are also available for each rack though ordered separately.