Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinematica Labstar Basic Lab Benchtop Autoclave

Catalog No. 08451075 Shop All Kinematica Products
Change view
Click to view available options
Capacity (Metric):
25 L
40 L
70 L
200 L
Dimensions (L x W x H):
450 x 520 x 660 mm
620 x 532 x 570 mm
850 x 700 x 825 mm
1250 x 750 x 900 mm
Weight (Metric):
45 kg
65 kg
144 kg
165 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Dimensions (L x W x H) Weight (Metric)
08-451-075 200 L 1250 x 750 x 900 mm 165 kg
08-451-073 40 L 620 x 532 x 570 mm 65 kg
08-451-074 70 L 850 x 700 x 825 mm 144 kg
08-451-072 25 L 450 x 520 x 660 mm 45 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 08-451-075 Supplier Kinematica Supplier No. A1280101
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kinematica Labstar Basic Compact Lab Benchtop Autoclave with angular chamber, touch screen, password protected. Recommended for the sterilization of simple materials or liquids without removal when dry.

  • Easy-to-use touchscreen
  • 10 variable program channels for maximum flexibility
  • Password-protected parameters
  • Round chamber (Labstar 200), Angular (Labstar 25, 40, 70)
  • Maximum allowable operating temperature up to 140°C and maximum pressure at 3.8 bar absolute
  • Equipped with electronic pressure sensor and PT100 temp sensor
  • Hinged door with locking mechanism powered by an electric motor
  • Sterilization chamber and door made with stainless steel
  • Housing made with brushed stainless steel

Specifications

Autoclavable Not Autoclavable
Amperage 200 amp, 3 Phase
Certifications/Compliance CE Compliant
Capacity (English) 53 gal.
Capacity (Metric) 200 L
Depth (Metric) Chamber 680 mm
Diameter (Metric) Chamber 500 mm
For Use With (Application) Steam Sterilization of Lab Equipment and Waste
For Use With (Equipment) Glass, Stainless Steel, Cloth, Plastic, Clothing, Biohazardous Materials, Agar, Media
Length (English) 49 in.
Height (Metric) Exterior 900 mm
Length (Metric) 1250 mm
Model Labstar
Product Type Autoclave
Temperature (English) 285°F
Temperature (Metric) 140°C
Weight (English) 365 lb.
Display Touch Display
Weight (Metric) 165 kg
Features Easy-To-Use Touchscreen, Password-Protected, 10 Variable Program Channels for Maximum Flexibility, Angular Chamber, Sterilization Chamber and Door Made of Stainless Steel 1.4404/AiSi316L, Equipped with Electronic Pressure Sensor and PT100 Temp Sensor, Hinged Door with Locking Mechanism Powered by an Electric Motor.
Includes Free Promotional Printer (Limited Time)
Frequency 60 Hz
Material Stainless Steel
Latex Free Yes
Operating Pressure 3.8 bar
Pressure (English) Feed Water External Plumbing
Programmability Programmable
Quantity 1 Ea.
Sterilization Method Steam Sterilization
Voltage 240 V
Width (English) Exterior 30 in.
Sterility Sterile
Width (Metric) Exterior 750 mm
Dimensions (L x W x H) 1250 x 750 x 900 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.